Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PlGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $532.55
Specifications
| Antigen | PLGF |
|---|---|
| Clone | 1038925 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB111991SP
![]() |
R&D Systems
MAB111991SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11199110
![]() |
R&D Systems
MAB111991100 |
100 μg |
Each for $532.55
|
|
|||||
Description
PLGF Monoclonal antibody specifically detects PLGF in Human samples. It is validated for nanSpecifications
| PLGF | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5228 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1038925 | |
| Unconjugated | |
| Mouse | |
| Human | |
| D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| CHO-derived human PlGF-2 protein, Leu19-Arg170, Accession # NP_002623 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title