Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PlGF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB2642
This item is not returnable.
View return policy
Description
PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Immunohistochemistry.Specifications
PLGF | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P49763 | |
PGF | |
E.coli-derived recombinant human PLGF Ala21-Ala149 (predicted) Accession # P49763 | |
100 μg | |
Primary | |
Detects human PLGF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CTGF, rhLDGF, rhPDGF, rhVEGF-C, rhVEGF-D, rhVEGF-B 186, rhVEGF 206, or recombinant mouse PlGF-2 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
358905 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5228 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction