Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PlGF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB264PB
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB264PB 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB264PB Supplier R&D Systems Supplier No. AB264PB
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PLGF Polyclonal specifically detects PLGF in Human samples. It is validated for Western Blot.

Specifications

Antigen PLGF
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. CAA38698
Gene Alias D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760
Gene Symbols PGF
Host Species Goat
Immunogen E. coli-derived recombinant human PlGF (R&D Systems, Catalog # 264-PG) Ala21-Arg149 Accession # CAA38698
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5228
Test Specificity Detects human PlGF but not VEGF in direct ELISAs and Western blots. This antibody will also recognize the heterodimer of VEGF and PlGF.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.