Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PLTP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5109SP
This item is not returnable.
View return policy
Description
PLTP Polyclonal specifically detects PLTP in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
PLTP | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
Sheep | |
Affinity Purified | |
RUO | |
5360 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Knockout Validated | |
P55058 | |
PLTP | |
Mouse myeloma cell line NS0-derived recombinant human PLTP isoform 1 Glu18-Val493 (Glu18Val) Accession # P55058 | |
25 μg | |
Primary | |
Detects human PLTP in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse PLTP is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction