Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PLUNC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1897
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1897 100 μg
AF1897SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1897 Supplier R&D Systems Supplier No. AF1897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PLUNC Polyclonal specifically detects PLUNC in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PLUNC
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, ELISA
Gene Accession No. Q9NP55
Gene Alias bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl
Gene Symbols BPIFA1
Host Species Goat
Immunogen E. coli-derived recombinant human PLUNC Gln20-Val256 Accession # Q9NP55
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51297
Test Specificity Detects human PLUNC in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.