Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Podocalyxin Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Catalog No. FAB1658G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1658G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1658G Supplier R&D Systems Supplier No. FAB1658G
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Podocalyxin Like
Applications Flow Cytometry
Classification Monoclonal
Clone 222328
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAB61574
Gene Alias GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL
Gene Symbols PODXL
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Podocalyxin Ser23-Arg427 Accession # AAB61574
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Test Specificity Detects human Podocalyxin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Podocalyxin or recombinant human Endoglycan is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.