Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Podocalyxin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1658
This item is not returnable.
View return policy
Description
Podocalyxin Like Polyclonal specifically detects Podocalyxin Like in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Podocalyxin Like | |
Polyclonal | |
Biotin | |
AAB61574 | |
PODXL | |
Mouse myeloma cell line NS0-derived recombinant human Podocalyxin Ser23-Arg427 Accession # AAB61574 | |
50 μg | |
Primary | |
Detects human Podocalyxin in Western blots. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL | |
GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL | |
Goat | |
Affinity Purified | |
RUO | |
5420 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction