Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Podoplanin Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF3670
This item is not returnable.
View return policy
Description
Podoplanin Polyclonal specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
Podoplanin | |
Polyclonal | |
Unconjugated | |
Q86YL7 | |
PDPN | |
Mouse myeloma cell line NS0-derived recombinant human Podoplanin Glu21-Lys123 Accession # Q86YL7 | |
100 μg | |
Primary | |
Detects human Podoplanin in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready | |
36-KD, aggrus, Gp36, Gp38, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, PDPN, podoplanin, RANDAM-2, T1A, T1A-2, T1-alpha | |
Sheep | |
Affinity Purified | |
RUO | |
10630 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction