Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Polypeptide GalNac Transferase 2/ GALNT2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7507
This item is not returnable.
View return policy
Description
GALNT2 Polyclonal specifically detects GALNT2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
GALNT2 | |
Polyclonal | |
Unconjugated | |
Q10471 | |
GALNT2 | |
Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 2/GALNT2 Lys52-Gln571 Accession # Q10471 | |
100 μg | |
Primary | |
Detects human Polypeptide GalNac Transferase 2/GALNT2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) GALNT1 and rhGALNTL-1 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
GalNAc-T2, GalNAc-T2EC 2.4.1.41, Polypeptide GalNAc transferase 2, polypeptide N-acetylgalactosaminyltransferase 2, pp-GaNTase 2, Protein-UDP acetylgalactosaminyltransferase 2, UDP-GalNAc transferase 2, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 2 (GalNAc-T2) | |
Sheep | |
Affinity Purified | |
RUO | |
2590 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction