Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Polypeptide GalNAc Transferase 3/GALNT3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7174
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7174 Supplier R&D Systems Supplier No. AF7174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Polypeptide GalNac Transferase 3/GALNT3 Polyclonal specifically detects Polypeptide GalNac Transferase 3/GALNT3 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Polypeptide GalNac Transferase 3/GALNT3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q14435
Gene Alias EC 2.4.1.41, GalNAc transferase 3, GalNAc-T3, GalNAc-T3DKFZp686C10199, HFTC, HFTCMGC61909, HHS, HHSPolypeptide GalNAc transferase 3, polypeptide GalNAc-transferase T3, polypeptide N-acetylgalactosaminyltransferase 3, pp-GaNTase 3, Protein-UDP acetylgalactosaminyltransferase 3, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 3 (GalNAc-T3)
Gene Symbols GALNT3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 3/GALNT3 Gln38-Asp633 Accession # Q14435
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2591
Test Specificity Detects human Polypeptide GalNac Transferase 3/GALNT3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) GALNT1 and rhGALNT4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.