Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PON2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4344SP
This item is not returnable.
View return policy
Description
PON2 Polyclonal specifically detects PON2 in Human samples. It is validated for Western Blot.Specifications
PON2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
A-esterase 2, Aromatic esterase 2, EC 3.1.1.2, EC 3.1.1.81, paraoxonase 2, paraoxonase nirs, PON 2, Serum aryldialkylphosphatase 2, serum paraoxonase/arylesterase 2 | |
Goat | |
Affinity Purified | |
RUO | |
5445 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Q15165 | |
PON2 | |
E. coli-derived recombinant human PON2 Ala30-Leu354 Accession # Q15165 | |
25 μg | |
Primary | |
Detects endogenous human PON2 in Western blots. In Western blots, this antibody shows no cross-reactivity with recombinant human PON1 or PON3. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction