Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PON2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4344
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4344 Supplier R&D Systems Supplier No. MAB4344
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PON2 Monoclonal specifically detects PON2 in Human samples. It is validated for Western Blot.

Specifications

Antigen PON2
Applications Western Blot
Classification Monoclonal
Clone 453709
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000296
Gene Alias A-esterase 2, Aromatic esterase 2, EC 3.1.1.2, EC 3.1.1.81, paraoxonase 2, paraoxonase nirs, PON 2, Serum aryldialkylphosphatase 2, serum paraoxonase/arylesterase 2
Gene Symbols PON2
Host Species Rat
Immunogen E. coli-derived recombinant human PON2 Ala30-Leu354 Accession # NP_000296
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5445
Test Specificity Detects endogenous human PON2 in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) PON1 or rhPON3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.