Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human POT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5299
This item is not returnable.
View return policy
Description
POT1 Monoclonal specifically detects POT1 in Human samples. It is validated for Western Blot.Specifications
POT1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q9NUX5 | |
POT1 | |
E. coli-derived recombinant human POT1 Met159-Pro316 Accession # Q9NUX5 | |
100 μg | |
Primary | |
Detects human POT1 in direct ELISAs. | |
Human | |
Purified |
Western Blot | |
688923 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
25913 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction