Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PP1 alpha Catalytic Subunit Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6105
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6105 Supplier R&D Systems Supplier No. MAB6105
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP1 alpha/PPP1A Monoclonal specifically detects PP1 alpha/PPP1A in Human samples. It is validated for Western Blot.

Specifications

Antigen PP1 alpha/PPP1A
Applications Western Blot
Classification Monoclonal
Clone 490314
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P62136
Gene Alias EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1A, PPP1AMGC1674, PPP1CA, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit
Gene Symbols PPP1CA
Host Species Mouse
Immunogen synthetic peptide corresponding to Gly304-Arg317 of human PP1 alpha catalytic subunit Gly304-Arg317 Accession # P62136
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5499
Test Specificity Detects human PP1 alpha Catalytic Subunit in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) PP1 beta or rhPP1 gamma is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.