Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PP1 Inhibitor-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4719SP
This item is not returnable.
View return policy
Description
PP1 Inhibitor-2 Monoclonal specifically detects PP1 Inhibitor-2 in Human samples. It is validated for Western Blot.Specifications
PP1 Inhibitor-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P41236 | |
PPP1R2 | |
E. coli-derived recombinant human PP1 Inhibitor-2 Met1-Ser205 Accession # P41236 | |
25 μg | |
Primary | |
Detects human PP1 Inhibitor-2 in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
488123 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IPP2, IPP-2, IPP2phosphatase inhibitor 2, MGC87148, phosphoprotein phosphatase, PP1 Inhibitor2, PPP1R2, protein phosphatase 1, regulatory (inhibitor) subunit 2, protein phosphatase inhibitor 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5504 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction