Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PP2C gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5595
This item is not returnable.
View return policy
Description
PP2C gamma/PPM1G Monoclonal specifically detects PP2C gamma/PPM1G in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PP2C gamma/PPM1G | |
Monoclonal | |
LYOPH | |
Western Blot 0.25 ug/mL, Immunohistochemistry 8-25 ug/mL | |
O15355 | |
PPM1G | |
E. coli-derived recombinant human PP2C gamma Met1-Asp546 Accession # O15355 | |
100 μg | |
Primary | |
Detects human PP2C gamma . | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
528322 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2Cgamma, PP2C-gamma, PPM1C, PPM1G, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5496 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction