Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PPAR alpha/NR1C1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH072300
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH072300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH072300 Supplier R&D Systems Supplier No. PPH072300
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen PPAR alpha/NR1C1
Applications ChIP Assay, ELISA, Gel Shift, Immunoprecipitation
Classification Monoclonal
Clone H0723
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Direct ELISA, Gel Supershift Assay, Chromatin Immunoprecipitation (ChIP)
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias alpha, hPPAR, NR1C1, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARA, PPARalpha, PPAR-alpha, PPARMGC2452
Gene Symbols PPARA
Host Species Mouse
Immunogen Recombinant human PPAR alpha/NR1C1 aa 4-96
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5465
Test Specificity This antibody specifically recognizes human PPAR alpha and cross-reacts with mouse PPAR alpha . This antibody does not cross-react with human PPAR gamma or PPAR delta . Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.