Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PPAR alpha/NR1C1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB24781SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB24781SP 25 μg
MAB24781100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB24781SP Supplier R&D Systems Supplier No. MAB24781SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen PPAR alpha/NR1C1
Applications Immunocytochemistry
Classification Monoclonal
Clone 2297B
Conjugate Unconjugated
Dilution Immunocytochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q07869
Gene Alias alpha, hPPAR, NR1C1, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARA, PPARalpha, PPAR-alpha, PPARMGC2452
Host Species Rabbit
Immunogen E. coli-derived recombinant human PPAR alpha/NR1C1 Met1-Asn99 Accession # Q07869
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 5465
Test Specificity Detects human PPAR alpha/NR1C1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.