Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PPAR delta/NR1C2 Antibody (Clone K9436), R&D Systems™
GREENER_CHOICE

Catalog No. PPK943600
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPK943600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPK943600 Supplier R&D Systems Supplier No. PPK943600
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

PPAR delta/NR1C2 Monoclonal specifically detects PPAR delta/NR1C2 in Human samples. It is validated for Western Blot, Gel Supershift Assay.

Specifications

Antigen PPAR delta/NR1C2
Applications Western Blot, Gel Shift
Classification Monoclonal
Clone K9436
Conjugate Unconjugated
Dilution Western Blot, Gel Supershift Assay
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias FAAR, MGC3931, NR1C2, NR1C2PPAR-delta, NUC1, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB, PPARD
Gene Symbols PPARD
Host Species Mouse
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5467
Target Species Human
Content And Storage Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.