Learn More
Human PPAR gamma/NR1C3 Antibody (Clone K8713), R&D Systems™

Description
Specifications
Specifications
| Antigen | PPAR gamma/NR1C3 |
| Applications | Western Blot, Immunoprecipitation, ELISA, Gel Shift |
| Classification | Monoclonal |
| Clone | K8713 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunoprecipitation, Direct ELISA, Gel Supershift Assay |
| Formulation | A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide |
| Gene Alias | CIMT1, NR1C3, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma |
| Gene Symbols | PPARG |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.