Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PPAR gamma/NR1C3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPA3409A00
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPA3409A00 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPA3409A00 Supplier R&D Systems Supplier No. PPA3409A00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPAR gamma/NR1C3 Monoclonal specifically detects PPAR gamma/NR1C3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Supershift Assay.

Specifications

Antigen PPAR gamma/NR1C3
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Shift
Classification Monoclonal
Clone A3409A
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Gel Supershift Assay
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias CIMT1, NR1C3, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Gene Symbols PPARG
Host Species Mouse
Immunogen Recombinant human PPAR gamma/NR1C3 Amino Acids 3-108
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5468
Test Specificity This antibody specifically recognizes human PPAR gamma 1 and gamma 2 and cross-reacts with mouse and rat PPAR gamma 1 and gamma 2. This antibody does not recognize human PPAR alpha and PPAR delta . Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20degrees C in a manual defrost freezer or at -70degrees C. Upon thawing, the antibody can be stored at 2degrees - 8degrees C for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.