Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PRAS40 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6408
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6408 Supplier R&D Systems Supplier No. AF6408
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PRAS40 Polyclonal specifically detects PRAS40 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen PRAS40
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q96B36
Gene Alias 40 kDa, AKT1 substrate 1 (proline-rich), AKT1S1, Lobe, MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate
Gene Symbols AKT1S1
Host Species Sheep
Immunogen E. coli-dervied recombinant human PRAS40 Gln119-Tyr256 Accession # Q96B36
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84335
Test Specificity Detects human PRAS40 in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.