Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PRDM14 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6175SP
This item is not returnable.
View return policy
Description
PRDM14 Polyclonal specifically detects PRDM14 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PRDM14 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC59730, PFM11, PR domain containing 14, PR domain zinc finger protein 14, PR domain-containing protein 14, PR-domain zinc finger protein 14 | |
Sheep | |
Affinity Purified | |
RUO | |
63978 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q9GZV8 | |
PRDM14 | |
E. coli-derived recombinant human PRDM14 Ala2-Asp201 Accession # Q9GZV8 | |
25 μg | |
Primary | |
Detects human PRDM14 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction