Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pref-1/DLK-1/FA1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1144
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Polyclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot.Specifications
Pref-1/DLK1/FA1 | |
Polyclonal | |
Biotin | |
AAA75364 | |
DLK1 | |
Mouse myeloma cell line NS0-derived recombinant human Pref-1 long isoform Ala24-Pro297 with Arg248Pro and Lys295Ser substitutions Accession # AAA75364 | |
50 μg | |
Primary | |
Detects human Pref-1/DLK-1/FA1 in Western blots. In this format, approximately 25% cross-reactivity with recombinant mouse Pref-1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
Goat | |
Affinity Purified | |
RUO | |
8788 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction