Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pregnancy Zone Protein/PZP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB82801100
This item is not returnable.
View return policy
Description
Pregnancy Zone Protein Monoclonal specifically detects Pregnancy Zone Protein in Human samples. It is validated for ELISA.Specifications
PregnancyZoneProtein | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6, CPAMD6, pregnancy zone protein, pregnancy-zone protein, PZP | |
Human embryonic kidney cell line HEK293-derived recombinant human Pregnancy Zone Protein/ PZP Thr26-Val1482 Accession # P20742 | |
100 μg | |
5858 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
ELISA | |
909855R | |
ELISA | |
P20742 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Pregnancy Zone Protein/PZP in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction