Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PRELP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB64471SP
This item is not returnable.
View return policy
Description
PRELP Monoclonal specifically detects PRELP in Human samples. It is validated for Neutralization.Specifications
PRELP | |
Monoclonal | |
LYOPH | |
Neutralization | |
P51888 | |
PRELP | |
Chinese hamster ovary cell line CHO-derived recombinant human PRELP Gln21-Ile382 Accession # P51888 | |
25 μg | |
Primary | |
Detects human PRELP in direct ELISA and Western blot. | |
Human | |
Purified |
Neutralization | |
754636 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
55 kDa leucine-rich repeat protein of articular cartilage, MST161, MSTP161, prolargin, prolargin proteoglycan, proline arginine-rich end leucine-rich repeat protein, proline/arginine-rich end leucine-rich repeat protein, Proline-arginine-rich end leucine-rich repeat protein, SLRR2A, SLRR2AMGC45323 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5549 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction