Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PRELP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6447
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6447 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6447 Supplier R&D Systems Supplier No. AF6447
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PRELP Polyclonal specifically detects PRELP in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.

Specifications

Antigen PRELP
Applications Immunocytochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunocytochemistry 5 to 15 μg/mL
Gene Accession No. P51888
Gene Alias 55 kDa leucine-rich repeat protein of articular cartilage, MST161, MSTP161, prolargin, prolargin proteoglycan, proline arginine-rich end leucine-rich repeat protein, proline/arginine-rich end leucine-rich repeat protein, Proline-arginine-rich end leucine-rich repeat protein, SLRR2A, SLRR2AMGC45323
Gene Symbols PRELP
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human PRELP Gln21-Ile382 Accession # P51888
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5549
Test Specificity Detects human PRELP in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.