Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Presenilin-1 C-Terminal Fragment Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF166SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF166SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF166SP Supplier R&D Systems Supplier No. AF166SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Presenilin-1 Polyclonal specifically detects Presenilin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.

Specifications

Antigen Presenilin-1
Applications Western Blot, Immunohistochemistry, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Presenilin1, Protein S182, PS-1, PS1presenilin-1, PSEN1, PSNL1, S182
Gene Symbols PSEN1
Host Species Goat
Immunogen E. coli-derived recombinant human Presenilin-1 aa 298-407
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5663
Test Specificity Detects human Presenilin-1 C-Terminal Fragment in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Presenilin-1 N-Terminal Fragment (aa 1-80), Presenilin-2 C-Terminal Fragment (aa 322-388), and rhPresenilin-2 N-Terminal Fragment (aa 1-80) is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.