Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Presenilin-1 N-Terminal Fragment Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB149
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB149 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB149 Supplier R&D Systems Supplier No. MAB149
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Presenilin-1 Monoclonal specifically detects Presenilin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Presenilin-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 121121
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49768
Gene Alias AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Presenilin1, Protein S182, PS-1, PS1presenilin-1, PSEN1, PSNL1, S182
Gene Symbols PSEN1
Host Species Mouse
Immunogen E. coli-derived recombinant human Presenilin-1 N-Terminal Fragment Lys1-Lys80 Accession # P49768
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5663
Test Specificity Detects human Presenilin-1 N-Terminal Fragment in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Presenilin-2 N-Terminal Fragment, rhPresenilin-1 C-Terminal Fragment, or rhPresenilin-2 C-Terminal Fragment.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.