Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PRG-1/LPPR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | PRG-1/LPPR4 |
---|---|
Clone | 368701 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
PRG-1/LPPR4 Monoclonal specifically detects PRG-1/LPPR4 in Human samples. It is validated for Western Blot.Specifications
PRG-1/LPPR4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
AAO85400 | |
9890 | |
E. coli-derived recombinant human PRG-1/LPPR4 Ser285-Asp715 Accession # AAO85400 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects PRG-1/LPPR4 in direct ELISAs and Western blots. |
368701 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LPPR4, LPPR4 lipid phosphate phosphatase-related protein type 4, LPR4, PHP1, PRG1, PRG-1, RP4-788L13.1 | |
LPPR4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title