Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate Angiopoietin-like Protein 4/ANGPTL4 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems BAF3485
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 4/ANGPTL4 Polyclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human, Primate samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Angiopoietin-like Protein 4/ANGPTL4 | |
Polyclonal | |
Biotin | |
Q9BY76 | |
ANGPTL4 | |
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 4, Leu165-Ser406, Accession # Q9BY76 | |
50 μg | |
Primary | |
Detects human ANGPTL4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) ANGPTL3, rhAngiopoietin-1, -2, -3, and -4 is observed.In Western blots, approximately 1% cross-reactivity with rhANGPLT4 N-terminal Fragment is observed. | |
Human, Primate | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
Goat | |
Affinity Purified | |
RUO | |
51129 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction