Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate CCL5/RANTES Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Catalog No. MAB678500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB678500 500 μg
MAB678SP 25 μg
MAB678100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB678500 Supplier R&D Systems Supplier No. MAB678500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 25 publications

CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Human, Primate samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Antigen CCL5/RANTES
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 21418
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.04-0.12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES
Gene Symbols CCL5
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL5/RANTES
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6352
Test Specificity Detects human and primate CCL5/RANTES in direct ELISAs and Western blots. When used in combination with the biotinylated anti-human/primate CCL5/RANTES detection antibody (Catalog # BAF278) in sandwich ELISAs, no significant cross-reactivity or interference is observed with recombinant human (rh) ANG, rhCNTF, rh beta-ECGF, rhEGF, mouse (m) EGF, rhEpo, bovine (b) FGF acidic, rhFGF acidic, bFGF basic, rhFGF basic, rhFGF-4, rhG-CSF, rhGM-CSF, recombinant mouse (rm) GM-CSF, rhGRO alpha , rhIFN-gamma , rhIGF-I, rhIGF-II, rhIL-1 alpha , rmIL-1 alpha , rhIL-1 beta , rmIL-1 beta , rhIL-1ra, recombinant rat (rr) IL-1ra, rhIL-2, rhIL-3, rmIL-3, rmIL-4, rhIL-5, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, rhIL-9, rmIL-9, rhIL-10, rhIL-11, rhLIF, rhMCP-1, rhM-CSF, rhMIP-1 alpha , rmMIP-1 alpha , rhMIP-1 beta , rmMIP-1 beta , rhOSM, human (h) PDGF, pPDGF, rhPDGF-AA, rhPDGF-AB, rhPDGF-BB, rhSCF, rmSCF, rhSLPI, rhTGF-alpha , pTGF-beta 1, hTGF-beta 1, rhTGF-beta 1, pTGF-beta 2, rhTGF-beta 2, recombinant canine TGF-beta 3, rhTGF-beta 3, recombinant amphibian TGF-beta 5, rhTNF-alpha , rmTNF-alpha , or rmTNF-beta .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.