Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CXCL1/GRO alpha/KC/CINC-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 20326R |
| Conjugate | Alexa Fluor 350 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.