Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate CXCL1/GRO alpha/KC/CINC-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC275T100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC275T100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC275T100UG Supplier R&D Systems Supplier No. IC275T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CXCL1/GRO alpha/KC/CINC-1
Applications Flow Cytometry
Classification Monoclonal
Clone 20326
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Gene Symbols CXCL1
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL1/GRO alpha/KC/CINC-1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2919
Test Specificity Detects human CXCL1/GRO alpha/KC/CINC-1 in ELISAs and Western blots. In Western blots, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) CXCL2/GRO beta and rhCXCL3/GRO gamma and no cross-reactivity with recombinant rat (rr)CINC-1, rrCINC-2 alpha , rrCINC-2 beta , rrCINC-3, rhMIP-1 alpha , recombinant mouse (rm)MIP-1 alpha , rmMIP-1 beta , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , or rmMIP-3 beta .
Target Species Human, Primate
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.