Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.50
Specifications
Antigen | CXCL1/GRO alpha/KC/CINC-1 |
---|---|
Clone | 20326R |
Dilution | Neutralization 0.600 - 9.00 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, Flow Cytometry, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB275RSP
![]() |
R&D Systems
MAB275RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB275R100
![]() |
R&D Systems
MAB275R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human, Primate samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).Specifications
CXCL1/GRO alpha/KC/CINC-1 | |
Neutralization 0.600 - 9.00 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
CXCL1 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
20326R | |
Neutralization, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human, Primate | |
2919 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Detects human and primate CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title