Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate GDF-15 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB957SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB957SP 25 μg
MAB957100 100 μg
MAB957500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB957SP Supplier R&D Systems Supplier No. MAB957SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

GDF-15 Monoclonal specifically detects GDF-15 in Human, Primate samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen GDF-15
Applications Western Blot, ELISA
Classification Monoclonal
Clone 147627
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99988
Gene Alias GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Gene Symbols GDF15
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human GDF-15 Ala197-Ile308 Accession # Q99988
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9518
Test Specificity Detects human GDF-15 in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human GDF-11, recombinant mouse (rm) GDF-5, rmGDF-6, rmGDF-7, or rmGDF-8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.