Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Primate IL-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB602SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB602SP 25 μg
MAB602100 100 μg
MAB602500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB602SP Supplier R&D Systems Supplier No. MAB602SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 14 publications

IL-2 Monoclonal specifically detects IL-2 in Human, Primate samples. It is validated for Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-2
Applications Immunocytochemistry, ELISA
Classification Monoclonal
Clone 5355
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_000577
Gene Alias aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Gene Symbols IL2
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-2 Ala21-Thr153 Accession # NP_000577
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Test Specificity Detects human and primate IL-2 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-3, rhIL-4, rhIL-6, rhIL-7, rhIL-8, recombinant mouse (rm) IL-1 beta , rmIL-2, rmIL-3, rmIL-4, rmIL-5, rmIL-6, or rmIL-7 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.