Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate MMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $549.50
Specifications
| Antigen | MMP-9 |
|---|---|
| Clone | 36020R |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB936RSP
![]() |
R&D Systems
MAB936RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB936R100
![]() |
R&D Systems
MAB936R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Primate samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| MMP-9 | |
| ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Primate | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
| Chinese hamster ovary cell line CHO-derived recombinant human/Primate MMP-9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MMP-9 in direct ELISAs. |
| 36020R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4318.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title