Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate MMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB936SP
This item is not returnable.
View return policy
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
MMP-9 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MMP9 | |
Chinese hamster ovary cell line CHO-derived recombinant human MMP-9 | |
25 μg | |
Primary | |
Detects human MMP-9 in Western blots. Does not react with the C-terminal truncated form (65 kDa) of recombinant human (rh) MMP-9. In Western blots, approximately 5% cross-reactivity with rhMMP-2 and no cross-reactivity with rhMMP-1 or rhMMP-3 is observed. | |
Human, Primate | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, ELISA | |
36020 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 3-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4318 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction