Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PRKD3/nPKC nu Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6478
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6478 Supplier R&D Systems Supplier No. AF6478
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

PRKD3/nPKC nu Polyclonal specifically detects PRKD3/nPKC nu in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen PRKD3/nPKC nu
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O94806
Gene Alias EC 2.7.11, EC 2.7.11.13, EPK2, EPK2PKD3, nPKC nu, nPKC-nu, PKC nu, PKC-NU, PRKCN, PRKCNnPKC-NU, Protein kinase C nu type, protein kinase C, nu, protein kinase D3, Protein kinase EPK2, protein-serine/threonine kinase, serine/threonine-protein kinase D3
Gene Symbols PRKD3
Host Species Sheep
Immunogen E. coli-derived recombinant human PRKD3/nPKC nu Ile465-Val559 Accession # O94806
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23683
Test Specificity Detects human PRKD3/nPKC nu in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.