Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro-BMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $655.00
Specifications
Antigen | BMP-2 |
---|---|
Clone | 253717 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
BMP-2 Monoclonal specifically detects BMP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BMP-2 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC | |
BMP2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
253717 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P12643 | |
650 | |
Mouse myeloma cell line NS0-derived recombinant human BMP-2 Ala22-Lys281 (Arg245Ser, Arg279Ser) Accession # P12643 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects the pro region of the human BMP-2 precursor in direct ELISAs and Western blots. This antibody does not recognize mature recombinant human BMP-2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title