Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pro-BNP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3604SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3604SP 25 μg
MAB3604 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3604SP Supplier R&D Systems Supplier No. MAB3604SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot.

Specifications

Antigen BNP
Applications Western Blot
Classification Monoclonal
Clone 370104
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P16860
Gene Alias BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB
Gene Symbols NPPB
Host Species Rat
Immunogen E. coli-derived recombinant human Pro-BNP His27-His134 Accession # P16860
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4879
Test Specificity Detects human Pro-BNP in direct ELISAs and Western blots. No cross-reactivity with recombinant human BNP (aa 103-134) is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.