Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro-BNP Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3604
This item is not returnable.
View return policy
Description
BNP Polyclonal specifically detects BNP in Human samples. It is validated for Western Blot.Specifications
BNP | |
Polyclonal | |
Unconjugated | |
P16860 | |
NPPB | |
E. coli-derived recombinant human Pro-BNP His27-His134 Accession # P16860 | |
100 μg | |
Primary | |
Detects human Pro-BNP in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
Goat | |
Affinity Purified | |
RUO | |
4879 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction