Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro Caspase-10 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6864
This item is not returnable.
View return policy
Description
Caspase-10/FLICE2 Polyclonal specifically detects Caspase-10/FLICE2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Caspase-10/FLICE2 | |
Polyclonal | |
Unconjugated | |
Q92851 | |
CASP10 | |
E. coli-derived recombinant human Pro Caspase-10 Met1-Pro228 Accession # Q92851 | |
100 μg | |
Primary | |
Detects human Pro Caspase-10 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Caspase-2, rhCaspase-3 (long and short forms), rhCaspase-3, rhCaspase-7, rhCaspase-8, rhCaspase-8 p10, rhCaspase-8 p20 and rhCaspase-9 is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Apoptotic protease Mch-4, CASP10, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, Caspase10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, Mch4, MCH4ALPS2 | |
Goat | |
Affinity Purified | |
RUO | |
843 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction