Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro- Collagen II Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $538.00
Specifications
Antigen | CollagenII |
---|---|
Clone | 819427R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3615SP
![]() |
R&D Systems
MAB3615SP |
25 μg |
Each for $241.00
|
|
|||||
MAB3615100
![]() |
R&D Systems
MAB3615100 |
100 μg |
Each for $538.00
|
|
|||||
Description
Collagen II Monoclonal specifically detects Collagen II in Human samples. It is validated for ELISA.Specifications
CollagenII | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P02458 | |
1280 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
819427R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, COL2A1, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC | |
Chinese hamster ovary cell line CHO-derived recombinant human Pro-Collagen II Gln26-Arg266 and Gly957-Leu1487 Accession # P02458 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Pro-Collagen II in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title