Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro-Corticoliberin/Pro-CRH Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5805
This item is not returnable.
View return policy
Description
Pro-Corticoliberin/Pro-CRH Polyclonal specifically detects Pro-Corticoliberin/Pro-CRH in Human samples. It is validated for Western Blot.Specifications
Pro-Corticoliberin/Pro-CRH | |
Polyclonal | |
Unconjugated | |
P06850 | |
CRH | |
E. coli-derived recombinant human Pro-Corticoliberin/Pro-CRH Leu25-Lys196 Accession # P06850 | |
100 μg | |
Primary | |
Detects human Pro-Corticoliberin/Pro-CRH in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CRF, CRH, CRH1, ProCorticoliberin | |
Sheep | |
Affinity Purified | |
RUO | |
1392 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction