Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pro-MMP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB907SP
This item is not returnable.
View return policy
Description
MMP-7 Monoclonal specifically detects MMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MMP-7 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P09237 | |
MMP7 | |
Human MMP-7 synthetic peptide AEYSLFPNSPKWTSKV, Ala93-Val108 Accession # P09237 | |
25 μg | |
Primary | |
Detects recombinant human (rh) Pro-MMP-7 (aa 18-267) in Western blots. In Western blots, no cross-reactivity with rhMMP-1, rhMMP-2, rhMMP-9 or mature rhMMP-7 (aa 95-267) is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
6A4 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
4316 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction