Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Procalcitonin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $585.00
Specifications
Antigen | Procalcitonin |
---|---|
Clone | 944002 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB83501SP
![]() |
R&D Systems
MAB83501SP |
25 μg |
Each for $214.00
|
|
|||||
MAB83501100
![]() |
R&D Systems
MAB83501100 |
100 μg |
Each for $585.00
|
|
|||||
Description
Procalcitonin Monoclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Procalcitonin | |
Western Blot 1 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P01258 | |
796 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
944002 | |
Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Human Procalcitonin synthetic peptide Ala26-Asp38 Accession # P01258 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Procalcitonin in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title