Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Procalcitonin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8350
This item is not returnable.
View return policy
Description
Procalcitonin Polyclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot.Specifications
Procalcitonin | |
Polyclonal | |
Western Blot 1 ug/mL | |
P01258 | |
CALCA | |
E. coli-derived recombinant human Procalcitonin Ala26-Gly117 (Asn57Asp) Accession # P01258 | |
100 μg | |
Primary | |
Detects human Procalcitonin in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Sheep | |
Affinity Purified | |
RUO | |
796 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction