Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Procalcitonin DuoSet ELISA
Click to view available options
Sufficient For:
Five 96-well plates
Description
The Human Procalcitonin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Procalcitonin.
Specifications
Specifications
Assay Range | 31.20 to 2,000pg/mL |
Conjugate | Biotin |
Immunoassay Kit Format | 96-well Strip Plate |
Product Type | Solid Phase Sandwich ELISA |
Sufficient For | Five 96-well plates |
For Use With (Application) | ELISA |
Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction